Learn More
Memmert™ Entry Port 23 mm for IF750, Standard Position Left Side Center/Center
Optional entry port for Memmert Incubators.
Brand: Memmert™ IF750F0
1418.57 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position left side center/center
Specifications
IF750 Incubator | |
Entry Port | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for IF750, Standard Position Left Side Center/Center > For Use With: IF750
Spot an opportunity for improvement?Share a Content Correction