Learn More
Notrax ESD Common Grounding Cord
Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment.
241.00 SEK - 317.00 SEK
Specifications
Product Type Grounding Cord
Color Black
Material Wire
View More Specs
Products 2
Product Code Brand Connector Types Price Quantity & Availability  
Product Code Brand Connector Types Price Quantity & Availability  
17711918
View Documents
Notrax
052SCG0100
Ring Connector
241.00 SEK
Each
Only null left
Estimated Shipment: 04-07-2025
Add to basket
17721918
View Documents
Notrax
052SCG0200
Female Connector
317.00 SEK
Each
Only null left
Estimated Shipment: 04-07-2025
Add to basket
Description
Description
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
  • 45 m cable connects wrist bands or bench mat to ground
  • Assures that all parts are at the same ground potential
  • Easy to install, simply snap it to a mat, plug in the wrist strap coil cord and connect terminal to ground
  • Resistor 1 Meg mat to ground
  • Available with a ring terminal or 10 mm female socket
Specifications
Specifications
Grounding Cord
Black
Wire
Documents
Product Certifications
Product Content Correction
Product Title
Notrax ESD Common Grounding Cord