Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
400.00 SEK - 1990.00 SEK
Product Code | Brand | Standard Taper Joint Side | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint Side | Price | Quantity & Availability | ||||||
11886104
|
Lenz Laborglasintrumente
03042949 |
NS 29/32 | N/A | |||||||
11953381
|
Lenz Laborglasintrumente
03041410 |
NS 14/23 |
1990.00 SEK
Pack of 10 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
||||||
Description
- Especially for rotary evaporators
- With center neck and conical joint
- Pear-shaped
- With inscription label
- Material: DURAN™
Specifications
Duran Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint