Learn More
Lenz Laborglasintrumente Extraction Thimbles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
438.00 SEK - 558.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10274051
|
Lenz Laborglasintrumente
05431203 |
250 mL |
558.00 SEK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||||
10243911
|
Lenz Laborglasintrumente
05431202 |
100 mL |
472.00 SEK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||||
10644392
|
Lenz Laborglasintrumente
05431201 |
70 mL |
438.00 SEK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||||
Description
- Extraction thimbles
- Glass insert with bores for solids
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Extraction Thimbles