Learn More
Lenz Laborglasintrumente Extractor Thimbles for Solids
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
864.00 SEK - 1350.00 SEK
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | |||||
10080221
|
Lenz Laborglasintrumente
01507145 |
NS 45/40 | NS 71/51 Socket |
1260.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10214101
|
Lenz Laborglasintrumente
01506045 |
NS 45/40 | NS 60/46 Socket |
989.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10355501
|
Lenz Laborglasintrumente
01506029 |
NS 29/32 | NS 60/46 Socket |
864.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10661892
|
Lenz Laborglasintrumente
01507129 |
NS 29/32 | NS 71/51 Socket |
1350.00 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
Description
- Expansion adapters
- With ground joints
- Acc. to DIN 12257
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Thimbles for Solids