Learn More
Wheaton™ Fine Tip LDPE Pasteur Pipette
Pasteur pipette for simple and safe liquid measurement and transfer.
861.00 SEK - 2315.00 SEK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Bulb Draw (Metric) | 1 mL |
Calibrated | No |
Certifications/Compliance | CE Marked |
Material | LDPE |
Product Code | Brand | Packaging Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Price | Quantity & Availability | ||||||
16414999
|
Wheaton™
711117F |
Bulk Bag |
2315.00 SEK
Pack of 3000 |
Estimated Shipment: 12-05-2026
Log in to see stock. |
||||||
16424999
|
Wheaton™
711117F10S |
10/Pk |
1830.00 SEK
Pack of 1500 |
Estimated Shipment: 20-10-2025
Log in to see stock. |
||||||
16434999
|
Wheaton™
711117F1S |
Individually Wrapped |
861.00 SEK
Pack of 500 |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
16444999
|
Wheaton™
711117F20S |
20/Pk |
1635.00 SEK
Pack of 1500 |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
Description
- For liquid transfer
- Designed to ensure safe sample transfer
Specifications
Not Autoclavable | |
No | |
LDPE | |
No | |
102 mm |
1 mL | |
CE Marked | |
20 ÎĽL | |
Bulb | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Wheaton™ Fine Tip LDPE Pasteur Pipette