Learn More
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Thermo Scientific Finger Tite Connectors for Thermo Scientific and Agilent Instruments offer easier fit, reliability and leak-free connections.
1085.00 SEK - 4570.00 SEK
Specifications
For Use With (Equipment) | TRACE 1300 Series GC and Agilent GC Instruments |
---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
10794827
|
Thermo Scientific™
290SA131 |
Split/Splitless and MS Starter Kit |
4570.00 SEK
Each |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
10197494
|
Thermo Scientific™
290SA132 |
Split/Splitless and FID Starter Kit |
4320.00 SEK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
10137524
|
Thermo Scientific™
290SA133 |
Split/Splitless Injector Base Seal |
1085.00 SEK
Pack of 2 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
Description
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent Instruments offer easier fit, reliability and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
- Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
- No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
- Easy to handle with the nut touchable even with a hot injector/detector
- No tools required
Specifications
TRACE 1300 Series GC and Agilent GC Instruments |