Learn More
Fisherbrand™ Immersion Baskets for Ultrasonic Cleaners
This immersion basket is designed specifically for the Fisherbrand™ 112xx Series Dual Frequency Ultrasonic Cleaners. The fine mesh makes this immersion basket ideal for small items. The extra-long handle makes handling safe and easy.
Brand:  Fisherbrand™ 2070380000
181.58 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10611432
  • 187.00 SEK / Each
Only null left
Estimated Shipment: 30-07-2025
Description
  • Fine mesh for small part security
  • Available in polypropylene or stainless steel
  • Extra-long handle for ease-of-use
Specifications
Specifications
Immersion Basket
FB11211 Ultrasonic Cleaning Unit
Polypropylene
74 x 50 mm
Videos
  • Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Fisherbrand™ Immersion Baskets for Ultrasonic Cleaners > Hostalen immersion basket; 74W x 50mm H