missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXN1 Mouse anti-Human, Clone: 1D8B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66337-1-IG-150UL
This item is not returnable.
View return policy
Description
FOXN1 is a 648 amino acid protein belonging to the winged helix/forkhead family It contains 1 fork-head DNA-binding domain and is a transcriptional regulator involved in development. FOXN1 is involved in directing lineage choices of multi-potential progenitor epithelial cells and controls important regulating genes of lympho-epithelial interactions. It modulates keratinocyte differentiation in hair follicles and nail apparatus and its defects lead to T-cell immunodeficiency congenital alopecia, nail dystrophy, macroscopic nudity and an inborn dysgenesis of the thymus.Specifications
FOXN1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15353 | |
FOXN1 | |
FOXN1 Fusion Protein Ag18207 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1D8B12 | |
Unconjugated | |
FOXN1 | |
FKHL20, forkhead box N1, Forkhead box protein N1, FOXN1, RONU, WHN | |
Mouse | |
Protein G | |
RUO | |
8456 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXN1 Mouse anti-Human, Clone: 1D8B12, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction