missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Galaxy™ 48 R Incubators

When a high level of cell culture isolation is required or very limited space is available in the lab, the stackable Galaxy 48R is the right choice. The high temperature disinfection, a seamless chamber and a fanless design makes it easy to clean and provides high contamination protection.

Brand:  Eppendorf™ CO48332001

Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 15723405

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Small footprint: can be used in smaller laboratories.
  • Large backlit display with on screen help menu.
  • Control and data logging of temperature, CO2 , door openings, and alarms.
  • Fanless design offers more usable space.
  • Six-sided direct heating system provides a uniform incubated environment.
  • Deep-drawn, easy-to-clean stainless steel chamber without seams or welds.
  • Low gas consumption.
  • Integrated alarm relay.
  • Available options based on model number:
    • O2 gas control.
    • Humidity package.
    • High temperature disinfection (4 hour at 120°C).
    • O2 control (e.g., for hypoxic studies).
    • 2-split inner glass door.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

International Power Supply, High Temperature Disinfection
Stainless Steel
1 to 19%
CO2 Incubator
48 L
IR Sensor
1 to 19% O2 Control
2-Split Inner Door
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Galaxy™ 48 R Incubators > 2-Split Inner Door, International Power Supply, High Temperature Disinfection, 1-19% O2 Control

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.