Learn More
GPR126 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17774-1-AP-150UL
Description
The GPR126 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GPR126. This antibody recognizes human, mouse, rat antigen. The GPR126 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
GPR126 | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Immunohistochemistry, ELISA | |
Unconjugated | |
150 μL | |
57211 | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.