Learn More
Sartorius™ Grade 292a High Purity Folded Filter Qualitative Filter Papers
Suitable for qualitative analyses that require low ash content
Brand: Sartorius™ FT-4-215-240
This item is not returnable.
View return policy
Description
- Made of pure cotton linters or cotton linters with refined pulp
- Medium dense, medium fast filtering
- Ash content <0.06% according to DIN 54370
- Supplied in folded format
Specifications
Folded | |
4 to 7 ÎĽm | |
Used for analytical purposes that require a low ash content | |
100/Pk. | |
Qualitative Filter Paper |
Cotton Linters, Refined Pulp | |
60ÎĽm/sec. | |
292a | |
0.19 mm | |
240 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Sartorius™ Grade 292a High Purity Folded Filter Qualitative Filter Papers > Diameter: 240 mm
Spot an opportunity for improvement?Share a Content Correction