Learn More
Sonics & Materials Half Wave Extender Probes
Recommended when working with tall, narrow vessels such as Erlenmeyer flasks
Brand: Sonics & Materials 630-0444
This item is not returnable.
View return policy
Description
- Screw into threaded end probes of identical diameter in place of the replaceable tip
- Fabricated from titanium alloy, Ti-6Al-4V
- Use a solid extender, not one with a replaceable tip, when processing samples containing organic solvents or low surface tension liquids
- Available on special order with threaded ends to accept replaceable tips
- Autoclavable
Specifications
Extender with Replaceable Tip | |
25 mm | |
Threaded End Probe |
1 in. | |
Titanium Alloy |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Sonics & Materials Half Wave Extender Probes > 25 mm
Spot an opportunity for improvement?Share a Content Correction