Learn More
Novus Biologicals™ Human Claudin-3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75328
This item is not returnable.
View return policy
Description
The Human Claudin-3 ELISA Kit (Colorimetric) measures human Claudin-3 in serum, plasma and other biological fluids.
Specifications
0.31 - 20 ng/mL | |
This kit recognizes Human CLDN3 in samples. No significant cross-reactivity or interference between Human CLDN3 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1365 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
0.19 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CLDN3 | |
1 Kit | |
Claudin-3 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Human Claudin-3 ELISA Kit (Colorimetric) > Quantity: 1 Kit; Host Species: Human
Spot an opportunity for improvement?Share a Content Correction