Learn More
Novus Biologicals™ Human Claudin-3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75328
Product Code. 18324051
  • 8310.00 SEK / 1 set
Description
The Human Claudin-3 ELISA Kit (Colorimetric) measures human Claudin-3 in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Human CLDN3 in samples. No significant cross-reactivity or interference between Human CLDN3 and analogues was observed.
Biological Fluids, Plasma, Serum
1365
Human
Storage of components varies. See protocol for specific instructions.
0.19 ng/mL
ELISA Kit (Colorimetric)
ELISA
CLDN3
1 Kit
Claudin-3
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Human Claudin-3 ELISA Kit (Colorimetric) > Quantity: 1 Kit; Host Species: Human