Learn More
Abnova™ Human COMMD1 Partial ORF (NP_689729, 91 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16132137
Abnova™ Human COMMD1 Partial ORF (NP_689729, 91 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16132137
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16132137
Brand: Abnova™ H00150684Q01.25ug
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for AP, Array, ELISA, WB-Re
COMMD1 is a regulator of copper homeostasis, sodium uptake, and NF-kappa-B (see MIM 164011) signaling (de Bie et al., 2005 [PubMed 16267171]).[supplied by OMIM]
Sequence: AAVISKFWKSHKTKIRESLMNQSRWNSGLRGLSWRVDGKSQSRHSAQIHTPVAIIELELGKYGQESEFLCLEFDEVKVNQILKTLSEVEESISTLISQPN
Specifications
Accession Number NP_689729
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 150684
Molecular Weight (g/mol) 36.74kDa
Name COMMD1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen AAVISKFWKSHKTKIRESLMNQSRWNSGLRGLSWRVDGKSQSRHSAQIHTPVAIIELELGKYGQESEFLCLEFDEVKVNQILKTLSEVEESISTLISQPN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C2orf5/MGC27155/MURR1
Common Name COMMD1
Gene Symbol COMMD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction
Product Title
Abnova™ Human COMMD1 Partial ORF (NP_689729, 91 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal >