missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human P2RX2 Full-length ORF (NP_777362.1, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16136802

Abnova™ Human P2RX2 Full-length ORF (NP_777362.1, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16136802
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16136802

Brand: Abnova™ H00022953P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel. Binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle. Six transcript variants encoding six distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MAAAQPKYPAGATARRLARGCWSALWDYETPKVIVVRNRRLGVLYRAVQLLILLYFVWYVFIVQKSYQESETGPESSIITKVKGITTSEHKVWDVEEYVKPPEGGSVFSIITRVEATHSQTQGTCPESIRVHNATCLSDADCVAGELDMLGNGLRTGRCVPYYQGPSKTCEVFGWCPVEDGASVSQFLGTMAPNFTILIKNSIHYPKFHFSKGNIADRTDGYLKRCTFHEASDLYCPIFKLGFIVEKAGESFTELAHKGGVIGVIINWDCDLDLPASECNPKYSFRRLDPKHVPASSGYNFRFAKYYKINGTTTRTLIKAYGIRIDVIVHGQAGKFSLIPTIINLATALTSVGVGSFLCDWILLTFMNKNKVYSHKKFDKMVDTPASEPAQASTPTDPKGLAQL

Specifications

Accession Number NP_777362.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 22953
Molecular Weight (g/mol) 71.2kDa
Name P2RX2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MAAAQPKYPAGATARRLARGCWSALWDYETPKVIVVRNRRLGVLYRAVQLLILLYFVWYVFIVQKSYQESETGPESSIITKVKGITTSEHKVWDVEEYVKPPEGGSVFSIITRVEATHSQTQGTCPESIRVHNATCLSDADCVAGELDMLGNGLRTGRCVPYYQGPSKTCEVFGWCPVEDGASVSQFLGTMAPNFTILIKNSIHYPKFHFSKGNIADRTDGYLKRCTFHEASDLYCPIFKLGFIVEKAGESFTELAHKGGVIGVIINWDCDLDLPASECNPKYSFRRLDPKHVPASSGYNFRFAKYYKINGTTTRTLIKAYGIRIDVIVHGQAGKFSLIPTIINLATALTSVGVGSFLCDWILLTFMNKNKVYSHKKFDKMVDTPASEPAQASTPTDPKGLAQL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC129601/P2X2
Common Name P2RX2
Gene Symbol P2RX2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human P2RX2 Full-length ORF (NP_777362.1, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.