Learn More
Abnova™ Human PCDHGB6 Full-length ORF (NP_115271.1, 1 a.a. - 820 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_115271.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56100 |
Molecular Weight (g/mol) | 116.2kDa |
Name | PCDHGB6 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | MGGSCAQRRRAGPRQVLFPLLLPLFYPTLSEPIRYSIPEELAKGSVVGNLAKDLGLSVLDVSARKLRVSAEKLHFSVDAESGDLLVKNRIDREQICKERRRCELQLEAVVENPLNIFHVIVVIEDVNDHAPQFDKKEIHLEIFESASAGTRLSLDPATDPDININSIKDYKINSNPYFSLMVRVNSDGGKYPELSLEKLLDREEQRSHSLILTALDGGDPPRSATAHIEISVKDTNDNPPVFSRDEYRISLSENLPPGSPVLQVTATDQDEGVNAEINYYFRSTAQSTKHMFSLDEKTGMIKNNQSFDFEDVERYTMEVEAKDGGGLSTQCKVIIEILDENDNSPEIIITSLSDQILENSPPGMVVALFKTRDLDFGGNGEVRCNIETDIPFKIYSSSNNYYKLVTDGALDREQTPEYNVTIVATDRGKPPLSSSRSITLYVADINDNAPVFDQTSYVVHVAENNPPGASIAQVSASDPDLGLNGHISYSIVASDLEPLAVSSYVSVSAQSGVVFAQRAFDHEQLRAFALTLQARDHGSPTLSANVSLRVLVGDRNDNAPRVLYPALGPDGSAFFDMVPRSAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLVFADNLQEILPDLSDRPVLSDPQAELQFYLVVALALISVLFLLAVILAIALRLRRSLSPATWDCFHPGLCVKSGPVVPPNYSEGTLPYSYNLCIAHTGTKEFNFLKCSVPLHSNEDMVCSVSPGALIPPHGGEDLTSHPETLTSVSFSFLCVIYLIVY |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.