Learn More
Abnova™ Human PHF2 Partial ORF (NP_005383, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a protein which contains a zinc finger-like PHD (plant homeodomain) finger, distinct from other classes of zinc finger motifs, and a hydrophobic and highly conserved domain. The PHD finger shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_005383 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5253 |
Molecular Weight (g/mol) | 36.63kDa |
Name | PHF2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 ug |
Immunogen | ATVPVYCVCRLPYDVTRFMIECDACKDWFHGSCVGVEEEEAPDIDIYHCPNCEKTHGKSTLKKKRTWHKHGPGQAPDVKPVQNGSQLFIKELRSRTFPS |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.