Learn More
Abnova™ Human SECISBP2 Partial ORF (NP_076982.3, 106 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The incorporation of selenocysteine into a protein requires the concerted action of an mRNA element called a sec insertion sequence (SECIS), a selenocysteine-specific translation elongation factor and a SECIS binding protein. With these elements in place, a UGA codon can be decoded as selenocysteine. The gene described in this record encodes a nuclear protein that functions as a SECIS binding protein. Mutations in this gene have been associated with a reduction in activity of a specific thyroxine deiodinase, a selenocysteine-containing enzyme, and abnormal thyroid hormone metabolism. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_076982.3 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79048 |
Molecular Weight (g/mol) | 36.74kDa |
Name | SECISBP2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | VPGSQYLYNQPSCYRGFQTVKHRNENTCPLPQEMKALFKKKTYDEKKTYDQQKFDSERADGTISSEIKSARGSHHLSIYAENSLKSDGYHKRTDRKSRII |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.