missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT43 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24338-1-AP-20UL
This item is not returnable.
View return policy
Description
IFT43, encoded by C14ORF179, is a subunit of the intraflagellar transport complex A (IFT-A) of primary cilia. The IFT-A is implicated in retrograde ciliary transport along axonemal microtubules. Defects in subunits of IFT-A has been associated with skeletal ciliopathies, including Sensenbrenner syndrome, Jeune syndrome, and Ellis-van-Creveld syndrome. Mutations in C14ORF179 has been linked to Sensenbrenner syndrome. (21378380).Specifications
IFT43 | |
Polyclonal | |
Unconjugated | |
IFT43 | |
C14orf179, IFT43 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112752 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96FT9 | |
IFT43 | |
IFT43 Fusion Protein Ag19434 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IFT43 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction