Learn More
Invitrogen™ BioPrime™ Total Genomic Labeling System
Complete genomic DNA labeling kits designed for use in array comparative genomic hybridization applications (aCGH) that allows for better call rates with precious samples
6410.00 SEK - 16440.00 SEK
Specifications
Detection Method | Fluorescence |
---|---|
Label or Dye | Alexa Fluor 3, Alexa Fluor 5 |
Final Product Type | Probes (Labeled DNA) |
Labeling Target | Genomic DNA |
Includes Label or Dye | Yes |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10343803
|
Invitrogen™
18097011 |
30 Reactions |
16440.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10109972
|
Invitrogen™
18097010 |
10 Reactions |
6410.00 SEK
30 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
These complete, all-inclusive kits are optimized to work across the widest range of sample input material with no need for pre-amplification. Highly concentrated exo-Klenow fragment of DNA polymerase I in a component limited reaction allow for consistent, robust DNA yields of ∽8μg with as little as 50ng of genomic DNA input. An optimized dye labeled nucleotide mix with new dNTP linkers, novel, application-specific Alexa Fluor 3 and 5 dyes, and improved buffer chemistry reduce labeling variation and increase signal to noise on arrays. Excitation and emission spectra of Alexa Fluor 3 and 5 dyes are suited for conventional two color scanners with no need to change settings. Complete kits are batch tested to assure quality in performance from lot to lot.
Contents and Storage:
The simplified kits contains Alexa Fluor 3 and Alexa Fluor 5 2x reaction mix, Exo-Klenow fragment (40U/μL), 5mM EDTA, TE Buffer, and control DNA (Salmon Sperm). The BioPrime Total Genomic Labeling System also contains PurelinkTM Genomic DNA purification columns and purification buffers (the BioPrime Total Genomic Labeling Module does not). New master mix formulation streamlines reaction set-ups, increasing consistency.
- Higher signal to noise with Alexa Fluor™ 3 and 5 dyes
- Less channel bias due to novel reaction formulation
- Widest range of input material (50ng - 3μg)
- Simplified workflow with master mix formulation
Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research
Specifications
Fluorescence | |
Probes (Labeled DNA) | |
Yes | |
BioPrime™ |
Alexa Fluor 3, Alexa Fluor 5 | |
Genomic DNA | |
Direct Labeling | |
Total Genomic Labeling System |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.