Learn More
Ionomycin, calcium salt, 100%, >98%, For HPLC analysis, MP Biomedicals™
2390.00 SEK - 9810.00 SEK
Chemical Identifiers
CAS | 56092-82-1 |
---|---|
Molecular Formula | C41H70CaO9 |
Molecular Weight (g/mol) | 747.08 |
MDL Number | MFCD00083634 |
InChI Key | RULWQOBWZOQCOY-SQNLDGQHSA-M |
PubChem CID | 71311740 |
IUPAC Name | calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoateShow More |
SMILES | [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)OShow More |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413272
|
MP Biomedicals
0215507001 |
1 mg |
2390.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423272
|
MP Biomedicals
0215507005 |
5 mg |
9810.00 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
56092-82-1 | |
747.08 | |
RULWQOBWZOQCOY-SQNLDGQHSA-M | |
calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoateShow More |
Specifications
Ionomycin Calcium Salt | |
56092-82-1 | |
1 | |
MFCD00083634 | |
[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O | |
747.08 | |
747.1 | |
Ionomycin, calcium salt, For HPLC analysis |
Crystalline Powder | |
White | |
C41H70CaO9 | |
RULWQOBWZOQCOY-SQNLDGQHSA-M | |
calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate | |
71311740 | |
>98% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ionomycin Calcium Salt
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Ionomycin Calcium Salt
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ionomycin, calcium salt, 100%, >98%, For HPLC analysis, MP Biomedicals™