Learn More
KIAA0513 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21478-1-AP-150UL
 View more versions of this product
Product Code. 16881065
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The function of this protein remains unknown.
Specifications
Specifications
KIAA0513
Polyclonal
Unconjugated
KIAA0513
KIAA0513
Rabbit
Antigen Affinity Chromatography
RUO
234797, 307907, 9764
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60268, Q8R0A7
KIAA0513, RGD1304884
KIAA0513 Fusion Protein Ag15624
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KIAA0513 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated