Learn More
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit
Applications: Flow Cytometry
4370.00 SEK - 10325.00 SEK
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | APC/Cy7 |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
Product Type | APC-Cy7 Antibody Labeling Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780304
|
Novus Biologicals™
765-0005 |
1 x 100 ÎĽg Reaction |
N/A
|
N/A | |||||
18731204
|
Novus Biologicals™
765-0015 |
1 mg |
N/A
|
N/A | |||||
18736893
|
Novus Biologicals™
765-0010 |
300 ÎĽg |
N/A
|
N/A | |||||
18759733
|
Novus Biologicals™
765-0030 |
3 x 0.01 mg Reaction |
N/A
|
N/A | |||||
Description
APC/Cy7 is a tandem conjugate. In the Lightning-Link APC/Cy7 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the APC in the form of long wavelength light at 776nm.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
APC/Cy7 | |
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent | |
APC-Cy7 Antibody Labeling Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit