missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LTA4H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13662-1-AP-20UL
This item is not returnable.
View return policy
Description
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.Specifications
LTA4H | |
Polyclonal | |
Unconjugated | |
LTA4H | |
Leukotriene A 4 hydrolase, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, LTA 4 hydrolase, LTA4, LTA4H | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16993, 299732, 4048 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09960, P24527, P30349 | |
LTA4H | |
LTA4H Fusion Protein Ag4568 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LTA4H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction