missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15474-1-AP-150UL
This item is not returnable.
View return policy
Description
MAGEA11 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita.Specifications
MAGEA11 | |
Polyclonal | |
Unconjugated | |
MAGEA11 | |
Cancer/testis antigen 1.11, CT1.11, MAGE 11, MAGE 11 antigen, MAGE11, MAGEA 11, MAGEA11, melanoma antigen family A, 11, Melanoma associated antigen 11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4110 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P43364 | |
MAGEA11 | |
MAGEA11 Fusion Protein Ag7778 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MAGEA11 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction