Learn More
Lenz Laborglasinstrumente™ Medium Shape Weighing Bottles
629.00 SEK - 1995.00 SEK
Specifications
Includes | Interchangeable NS stopper, inscription label |
---|---|
Product Line | Bottle |
Product Type | Weighing Bottle |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
11798104
|
Lenz Laborglasinstrumente™
05565550 |
48 mm |
1995.00 SEK
Pack of 6 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
11788104
|
Lenz Laborglasinstrumente™
05565534 |
30 mm |
1420.00 SEK
Pack of 6 |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
10030181
|
Lenz Laborglasinstrumente™
05565540 |
38 mm |
1675.00 SEK
Pack of 6 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
11886164
|
Lenz Laborglasinstrumente™
05565580 |
75 mm |
629.00 SEK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
Specifications
Interchangeable NS stopper, inscription label | |
Bottle | |
Weighing Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Medium Shape Weighing Bottles