Learn More
Mitomycin C, MP Biomedicals™
The most widely used Mitomycin antibiotic. Mitomycin C, MP Biomedicals is the least toxic of the mitomycin antibiotics, with the most potent antitumor activity. It is an inhibitor of DNA synthesis, nuclear division, and cancer cells.
1390.00 SEK - 8720.00 SEK
Chemical Identifiers
CAS | 50-07-7 |
---|---|
Molecular Formula | C15H18N4O5 |
Molecular Weight (g/mol) | 334.332 |
InChI Key | NWIBSHFKIJFRCO-WUDYKRTCSA-N |
Synonym | Ametycin, 7-Amino-9-a-methoxymitosane, Mit-C, Mitocin-C, Mitomycinum, Mutamycin, Azirizino[2', 3': 3,4] pyrrolo [1,2-a] indole 4,7-dione-6-amino-1,1a, 2, 8, 8a, 8b-hexahydro- 8- (hydroxymethyl) 8a-methoxy-5-methyl carbamateShow More |
PubChem CID | 5746 |
ChEBI | CHEBI:27504 |
SMILES | CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17670867
|
MP Biomedicals
0210049802 |
2 mg |
1390.00 SEK
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17680867
|
MP Biomedicals
0210049810 |
10 mg |
4645.00 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17690867
|
MP Biomedicals
0210049820 |
20 mg |
8720.00 SEK
20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Antibacterial to gram positive, gram negative and acid-fast bacilli
- Decomposes DNA in cancer cells by forming a complex with DNA
- Inhibits division of cancer cells by interfering with the biosynthesis of DNA
Chemical Identifiers
50-07-7 | |
334.332 | |
Ametycin, 7-Amino-9-a-methoxymitosane, Mit-C, Mitocin-C, Mitomycinum, Mutamycin, Azirizino[2', 3': 3,4] pyrrolo [1,2-a] indole 4,7-dione-6-amino-1,1a, 2, 8, 8a, 8b-hexahydro- 8- (hydroxymethyl) 8a-methoxy-5-methyl carbamateShow More | |
CHEBI:27504 |
C15H18N4O5 | |
NWIBSHFKIJFRCO-WUDYKRTCSA-N | |
5746 | |
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N |
Specifications
Powder | |
6 to 9 (Stability) | |
850ÎĽg/mg minimum | |
Ametycin, 7-Amino-9-a-methoxymitosane, Mit-C, Mitocin-C, Mitomycinum, Mutamycin, Azirizino[2', 3': 3,4] pyrrolo [1,2-a] indole 4,7-dione-6-amino-1,1a, 2, 8, 8a, 8b-hexahydro- 8- (hydroxymethyl) 8a-methoxy-5-methyl carbamate | |
NWIBSHFKIJFRCO-WUDYKRTCSA-N | |
334.332 | |
CHEBI:27504 | |
Antibiotic |
50-07-7 | |
EmM(367 nm) = 21.84 (water); EmM(357 nm) = 22.78 (methanol); E1% (216 nm) = 742 (methanol); E1% (360 nm) = 742 (methanol); E1% (560 nm) = 0.06 (methanol) (Lit.) | |
C15H18N4O5 | |
Soluble in dilute aqueous acids or dilute aqueous bases. | |
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N | |
5746 | |
334.33 |
Safety and Handling
Recommended Storage : Store at +4°C.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Mitomycin C, Antibiotic Class
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Carcinogenicity Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H351-Suspected of causing cancer.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Mitomycin
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Mitomycin C, MP Biomedicals™