missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ alpha-Chymotrypsin from Bovine Pancreas
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains on carboxyl end of the bond
477.00 SEK - 7240.00 SEK
Specifications
Concentration | 1X |
---|---|
Color | White |
For Use With (Application) | Catalyzes the hydrolysis of Peptide bonds involving L-isomers of Tyrosine, Phenylalanine, Tryptophan |
Content And Storage | 0°C |
Source | Bovine pancreas |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438521
|
MP Biomedicals™
02152272.2 |
250 mg |
477.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448521
|
MP Biomedicals™
0215227201 |
1 g |
1295.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458521
|
MP Biomedicals™
0215227205 |
5 g |
7240.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
α-Chymotrypsin preferentially catalyzes the hydrolysis of peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan.
- CAS Number: 9004-07-3
- EC Number: 232-671-2
- MDL Number: MFCD00130481
- Extinction Coefficient: 20.4
- Format: white powder
- From Bovine Pancreas
- Crystallized, dialyzed, salt-free lyophilized powder
Specifications
1X | |
Catalyzes the hydrolysis of Peptide bonds involving L-isomers of Tyrosine, Phenylalanine, Tryptophan | |
Bovine pancreas | |
25 | |
Crystallized, Lyophilized |
White | |
0°C | |
alpha-Chymotrypsin | |
Heavy Metals, natural Trypsin inhibitors to various degrees, Organophosphorus compounds, AEBSF, alpha-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and a-2-Macroglobulin |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Alpha-Chymotrypsin, 1Ă— Crystallized
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Chymotrypsin
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ alpha-Chymotrypsin from Bovine Pancreas