missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYST2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13751-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the hepatoma-derived growth factor family. The encoded protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation. This gene was thought initially to be located on chromosome X, however, that location has been determined to correspond to a related pseudogene. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
MYST2 | |
Polyclonal | |
Unconjugated | |
KAT7 | |
HBO1, HBOA, KAT7, MYST 2, MYST2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11143, 217127, 303470 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95251, Q5SVQ0, Q810T5 | |
Kat7 | |
MYST2 Fusion Protein Ag4693 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MYST2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction