Learn More
Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers
Store and transport dry and solid materials, specimens and general lab supplies in these stackable containers which feature positive-lock lids that form a tight seal.
584.09 SEK - 5928.71 SEK
Specifications
Material | HDPE |
---|---|
Certifications/Compliance | NSF approved for food and beverage storage. |
Graduations | Molded |
Includes | Lid |
No. per Pack | 1 Ea. |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
12726737
|
Thermo Scientific™
7142-0015 |
15 L | 30.5 cm | 24 cm |
5928.71 SEK
Pack of 6 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||
10204311
|
Thermo Scientific™
7142-0020 |
20 L | 30.5 cm | 38 cm |
1272.13 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
10070631
|
Thermo Scientific™
7142-0015 |
15 L | 30.5 cm | 24 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Store and transport dry and solid materials, specimens and general lab supplies using Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers. Containers are stackable and have positive-lock lids that seal tight for storing both dry and solid materials.
Features:- High-density, polyethylene containers provide excellent chemical and temperature resistance
- Easy-grip, molded-in handles are comfortable and durable for lifting and moving
- Positive-lock lid provides a tight seal for transport or storage
- National Sanitation Foundation (NSF) approved for food and beverage storage
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
HDPE | |
Molded | |
1 Ea. |
NSF approved for food and beverage storage. | |
Lid |