Learn More
NFKBIE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11273-1-AP-20UL
This item is not returnable.
View return policy
Description
Inhibits NF-kappa-B by complexing with and trapping it in the cytoplasm. Inhibits DNA-binding of NF-kappa-B p50-p65 and p50-c-Rel complexes.Specifications
NFKBIE | |
Polyclonal | |
Unconjugated | |
NFKBIE | |
I kappa B epsilon, IkappaBepsilon, IkB E, IkB epsilon, IKBE, NF kappa B inhibitor epsilon, NF kappa BIE, NFKBIE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18037, 316241, 4794 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00221, O54910 | |
Nfkbie | |
NFKBIE Fusion Protein Ag1800 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Product Title
NFKBIE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction