Learn More
NOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17772-1-AP-20UL
 View more versions of this product
Product Code. 16877744
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Three splice variants of NOX1 have been identified, NOH-1L, NOH-1S and NOH-1Lv. NOH-1S is a voltage-gated proton channel that participates in the regulation of cellular pH and is blocked by zinc. NOH-1L is a pyridine nucleotide-dependent oxidoreductase that generates superoxide and might conduct H (+) ions as part of its electron transport mechanism, whereas NOH-1S does not contain an electron transport chain. NOX1 have the potential to be effective treatments for a range of ischemic diseases.
Specifications
Specifications
NOX1
Polyclonal
Unconjugated
NOX1
GP91 2, Mitogenic oxidase 1, MOX 1, MOX1, NADPH oxidase 1, NOH 1, NOH1, NOX 1, NOX1
Rabbit
Antigen Affinity Chromatography
RUO
114243, 237038, 27035
-20°C
Liquid
Western Blot, ELISA
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8CIZ9, Q9WV87, Q9Y5S8
Nox1
NOX1 Fusion Protein Ag12187
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated