Learn More
Corning™ PETG Baffled Erlenmeyer Flask with Vent Cap
BIOPHARMAGREENER_CHOICE
Constructed of optically clear polyethylene terephthalate (PETG) and ideal for shaker culture applications.
Brand:  Corning™ 431285
 View more versions of this product
Product Code. 15803885
  • 6280.00 SEK / Pack of 4
Only null left
Estimated Shipment: 06-11-2025
Description
The Corning™ Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are ready-to-use, ideal for shaker culture applications, and storage.
  • Available with baffled bottom
  • Molded-in graduations for accuracy
  • Large vent cap for continuous gas exchange while preventing leakage
  • Highest Sterility Assurance Level (SAL) of 10−6
  • Nonpyrogenic and RNase-/DNase-free
  • Made from optically-clear polyethylene terephthalate(PTEG)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nonpyrogenic, DNase/RNase-free
Polypropylene
4/Cs.
Fernbach
Vented
Orange
5 L
1/Pk.
Sterile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number34424001 Certificate TypeCertificate of Quality Date21/06/2025 Catalog Number431285
Lot Number15722001 Certificate TypeCertificate of Quality Date04/12/2024 Catalog Number431285
Promotions
Promotions
Product Content Correction
Product Title
Corning™ PETG Baffled Erlenmeyer Flask with Vent Cap > Baffled; 5 L; None
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.