Learn More
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
CAS: 9003-05-8 | (C3H5NO)n | 72.06 g/mol
423.00 SEK - 5835.00 SEK
Chemical Identifiers
CAS | 9003-05-8, 64-17-5 |
---|---|
Molecular Formula | (C3H5NO)n |
MDL Number | MFCD00084392 |
InChI Key | HRPVXLWXLXDGHG-UHFFFAOYSA-N |
SMILES | NC(=O)C(-*)C-* |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10357740
|
Thermo Scientific Acros
178040050 |
5 g | Glass bottle |
423.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10675432
|
Thermo Scientific Acros
178042500 |
250 g | Glass bottle |
3770.00 SEK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174430
|
Thermo Scientific Acros
178045000 |
500 g | Glass bottle |
5835.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9003-05-8, 64-17-5 | |
MFCD00084392 | |
NC(=O)C(-*)C-* |
(C3H5NO)n | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N |
Specifications
White to Yellow | |
Granules | |
[-CH2CH(CONH2)-]n | |
1.3000g/mL | |
Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, glycerol, lactic acid | |
NC(=O)C(-*)C-* | |
71.08 | |
CHEBI:28619 | |
Authentic | |
1.3 |
9003-05-8, 64-17-5 | |
(C3H5NO)n | |
MFCD00084392 | |
acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
5 to 6,000,000 (Average ) | |
13% max. | |
Poly(acrylamide) |
Safety and Handling
missing translation for 'rtecsNumber' : AS3700000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.