missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55148-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to PPP3CB.
PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.Specifications
PPP3CB | |
Polyclonal | |
Unconjugated | |
PPP3CB | |
CALNA2, CALNB, CAM PRP catalytic subunit, CNA2, PPP3CB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19056, 24675, 5532 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16298, P20651, P48453 | |
Ppp3cb | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPP3CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction