Learn More
PSME4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18799-1-AP-150UL
 View more versions of this product
Product Code. 16888634
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The antibody is expected to recognize all 4 isoforms of PSME4 but appears to mainly detect isoform 2 of 140 kDa.
proteasome (prosome, macropain) activator subunit 4 (PSME4) is subunit of the proteasome. PSME4 has been found to activate proteasomal cleavage of peptides in an energy-independent manner and may be involved in spermatogenesis. PSME4 may also be involved in DNA repair.
Specifications
Specifications
PSME4
Polyclonal
Unconjugated
PSME4
KIAA0077, PA200, Proteasome activator PA200, PSME4
Rabbit
Antigen Affinity Chromatography
RUO
103554, 23198, 498433
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14997, Q5SSW2
Psme4
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PSME4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated