Learn More
Pyrex™ PBT Screwcaps
Use with reagent bottles, culture tubes and other screwtop bottles. Pyrex™ PBT Screwcaps come in a wide range of sizes for your laboratory needs. Sealing discs provide inert inner seal, resistance to moisture vapor, and are great for culture work.
615.00 SEK - 1225.00 SEK
Specifications
Description | Screw cap |
---|---|
Includes | PTFE faced rubber insert |
Material | PBT |
Autoclavable | Autoclavable |
Closure Type | Screw Cap |
Product Code | Brand | Thread Size (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||||
10300025
|
Pyrex
4503/13 |
13 mm | Culture tubes |
615.00 SEK
Pack of 20 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
10165324
|
Pyrex
4503/15 |
15 mm | Culture tubes |
662.00 SEK
Pack of 20 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
10399974
|
Pyrex
4503/20 |
20 mm | Reagent and media bottles, culture tubes |
795.00 SEK
Pack of 20 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
10389974
|
Pyrex
4503/22 |
22 mm | Culture tubes |
831.00 SEK
Pack of 20 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
10034794
|
Pyrex
4503/25 |
25 mm | Reagent and media bottles, culture tubes |
964.00 SEK
Pack of 20 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
15985410
|
Pyrex
4503/30 |
30 mm | Culture tubes |
1225.00 SEK
Pack of 20 |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||||
12579835
|
Pyrex
4503/30 |
30 mm | Culture tubes |
1225.00 SEK
Pack of 20 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
|||||
10181930
|
Pyrex
4503/42 |
42 mm | Reagent bottles |
1145.00 SEK
Pack of 10 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
Description
- PTFE faced sealing disc for chemical resistance
- Withstands hot air sterilization to 180deg.C
- Autoclavable
Specifications
Screw cap | |
PBT | |
Screw Cap |
PTFE faced rubber insert | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Pyrex™ PBT Screwcaps