Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Designed to support Raschig rings within plain fractionating columns.
1025.00 SEK - 1480.00 SEK
Specifications
For Use With (Application) | Designed to support Raschig rings within plain fractionating columns |
---|---|
Type | Column packing support |
Material | Pyrex Borosilicate Glass |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15557800
|
Quickfit
EX13/12 |
19/26 socket size |
1025.00 SEK
Pack of 5 |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15567800
|
Quickfit
EX13/25 |
34/35 socket size |
1480.00 SEK
Pack of 5 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15577800
|
Quickfit
EX13/26 |
40/38 socket size |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Support is passed through socket of column and rests at the constriction above the cone
- Manufactured from chemically resistant borosilicate glass
Specifications
Designed to support Raschig rings within plain fractionating columns | |
Column packing support | |
Pyrex Borosilicate Glass |