missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD51AP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11255-1-AP-20UL
This item is not returnable.
View return policy
Description
May participate in a common DNA damage response pathway associated with the activation of homologous recombination and double-strand break repair. Functionally cooperates with PALB2 in promoting of D-loop formation by RAD51. Binds to single and double stranded DNA, and is capable of aggregating DNA. Also binds RNA.Specifications
RAD51AP1 | |
Polyclonal | |
Unconjugated | |
RAD51AP1 | |
PIR51, RAD51 associated protein 1, RAD51 interacting protein, RAD51AP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10635, 19362, 689055 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C551, Q96B01 | |
RAD51AP1 | |
RAD51AP1 Fusion Protein Ag1781 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RAD51AP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction