missing translation for 'onlineSavingsMsg'
Learn More
STEMCELL Technologies Rat Recombinant GM-CSF Product Code.: 17139361

STEMCELL Technologies Rat Recombinant GM-CSF

Product Code. p-7217551
100 μg, 100µg
Encompass_Preferred
Click to view available options
Quantity:
100 μg
1000 μg
20 μg
Unit Size:
1000µg
100µg
20µg
This item is not returnable. View return policy

Product Code. 17139361

Brand: STEMCELL Technologies 78018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Granulocyte-macrophage colony-stimulating factor

  • Granulocyte-macrophage colony-stimulating factor (GM-CSF) promotes the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils, eosinophils, and macrophages
  • In synergy with other cytokines such as stem cell factor, IL-3, erythropoietin, and thrombopoietin, it also stimulates erythroid and megakaryocyte progenitor cells
  • GM-CSF is produced by multiple cell types, including stromal cells, Paneth cells, macrophages, dendritic cells (DCs), endothelial cells, smooth muscle cells, fibroblasts, chondrocytes, and Th1 and Th17 cells T cells
  • The receptor for GM-CSF (GM-CSFR) is composed of two subunits: the cytokine-specific α subunit (GMRα; CD116) and the common subunit βc (CD131) shared with IL-3 and IL-5 receptors

Specifications

Name Colony-stimulating factor 2, CSF-2, MGI-1GM, Pluripoietic-alpha
Quantity 100 μg
Cell Type Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Monocytes; Myeloid Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies Rat Recombinant GM-CSF

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.