missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 30S ribosomal protein S11 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S11, was expressed in the hosts and tags indicated.

3180.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_417756.1
Gene ID (Entrez) 31655987
Name 30S ribosomal protein S11 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16025621
View Documents
enQuireBio™
QP8621-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16065621
View Documents
enQuireBio™
QP8621-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16015621
View Documents
enQuireBio™
QP8621-ec-100ug
100 ÎĽg
11590.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16045621
View Documents
enQuireBio™
QP8621-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16055621
View Documents
enQuireBio™
QP8621-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16035621
View Documents
enQuireBio™
QP8621-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_417756.1
30S ribosomal protein S11 Protein
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
31655987
Research Use Only
rpsK
Recombinant Protein
E. coli
AKAPIRARKR VRKQVSDGVA HIHASFNNTI VTITDRQGNA LGWATAGGSG FRGSRKSTPF AAQVAAERCA DAVKEYGIKN LEVMVKGPGP GRESTIRALN AAGFRITNIT DVTPIPHNGC RPPKKRRV
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli 30S ribosomal protein S11 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.