missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant E. coli Aerobic glycerol-3-phosphate dehydrogenase Protein Product Code.: 16057190

enQuireBio™ Recombinant E. coli Aerobic glycerol-3-phosphate dehydrogenase Protein

Product Code. 16057190
200 μg, 200µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16057190

Brand: enQuireBio™ QP7047ye200ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) Aerobic glycerol-3-phosphate dehydrogenase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_417884.1
Gene ID (Entrez) 947934
Name Aerobic glycerol-3-phosphate dehydrogenase Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol glpD
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species Yeast
Protein Tag His
Sequence METKDLIVIG GGINGAGIAA DAAGRGLSVL MLEAQDLACA TSSASSKLIH GGLRYLEHYE FRLVSEALAE REVLLKMAPH IAFPMRFRLP HRPHLRPAWM IRIGLFMYDH LGKRTSLPGS TGLRFGANSV LKPEIKRGFE YSDCWVDDAR LVLANAQMVV RKGGEVLTRT RATSARRENG LWIVEAEDID TGKKYSWQAR GLVNATGPWV KQFFDDGMHL PSPYGIRLIK GSHIVVPRVH TQKQAYILQN EDKRIVFVIP WMDEFSIIGT TDVEYKGDPK AVKIEESEIN YLLNVYNTHF KKQLSRDDIV WTYSGVRPLC DDESDSPQAI TRDYTLDIHD ENGKAPLLSV FGGKLTTYRK LAEHALEKLT PYYQGIGPAW TKESVLPGGA IEGDRDDYAA RLRRRYPFLT ESLARHYART YGSNSELLLG NAGTVSDLGE DFGHEFYEAE LKYLVDHEWV RRADDALWRR TKQGMWLNAD QQSRVSQWLV EYTQQRLSLA S
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Aerobic glycerol-3-phosphate dehydrogenase Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.