missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Parathyroid Hormone Protein

A DNA sequence encoding the Escherichia coli (strain K12) Parathyroid Hormone, was expressed in the hosts and tags indicated.

3180.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_415722.1
Gene ID (Entrez) 945765
Name Parathyroid Hormone Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16018170
View Documents
enQuireBio™
QP7318-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16058170
View Documents
enQuireBio™
QP7318-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16008170
View Documents
enQuireBio™
QP7318-ec-100ug
100 ÎĽg
11590.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16038170
View Documents
enQuireBio™
QP7318-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16048170
View Documents
enQuireBio™
QP7318-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028170
View Documents
enQuireBio™
QP7318-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_415722.1
Parathyroid Hormone Protein
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
945765
Research Use Only
PTH
Recombinant Protein
E. coli
MTIKLIVGLA NPGAEYAATR HNAGAWFVDL LAERLRAPLR EEAKFFGYTS RVTLGGEDVR LLVPTTFMNL SGKAVAAMAS FFRINPDEIL VAHDELDLPP GVAKFKLGGG HGGHNGLKDI ISKLGNNPNF HRLRIGIGHP GDKNKVVGFV LGKPPVSEQK LIDEAIDEAA RCTEMWFTDG LTKATNRLHA FKAQ
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Parathyroid Hormone Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.