missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-2 gp-36 397aa Protein
A cDNA sequence encoding the HIV-2 gp-36 397aa was constructed and used to recombinantly synthesize the protein.
2800.00 SEK - 20560.00 SEK
Specifications
Name | HIV-2 gp-36 397aa Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989888
|
enQuireBio™
QP12270-100ug |
100 ÎĽg |
2800.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15909898
|
enQuireBio™
QP12270-500ug |
500 ÎĽg |
10695.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15999888
|
enQuireBio™
QP12270-1mg |
1 mg |
20560.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
HIV-2 gp-36 397aa Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
0.01M Na2CO3, 0.01M Na3EDTA, 0.014 Mb-mercaptoethanol, 0.05% Tween-20. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
EQTMVQDDPSTCRGEFLYCNMTWFLNWIENKTHRNYAPCHIKQIINTWHKVGRNVYLPPREGELSCNSTVTSIIANIDWQNNNQTNITFSAEVAELYRLELGDYKLVEITPIGFAPTKEKRYSSAHGRHTRGVFVLGFLGFLATAGSAMGAASLTVSAQSRTLLAGIVQQQQQLLDVVKRQQELLRLTVWGTKNLQARVTAIEKYLQDQARLNSWGCAFRQVCHTTVPWVNDSLAPDWDNMTWQEWEKQVRYLEANISKSLEQAQIQQEKNMYELQKLNSWDIFGNWFDLTSWVKYIQYGVLIIVAVIALRIVIYVVQMLSRLRKGYRPVFSSPPGYIQQIHIHKDRGQPANEETEEDGGSNGGDRYWPWPIAYIHFLIRQLIRLLTRLYSICSQAC | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant HIV-2 gp-36 397aa Protein