missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human mRNA-decapping enzyme 1A Protein Product Code.: 16063241

enQuireBio™ Recombinant Human mRNA-decapping enzyme 1A Protein

Product Code. 16063241
500 μg, 500µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16063241

Brand: enQuireBio™ QP8201ec500ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) mRNA-decapping enzyme 1A, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001277133.1
Gene ID (Entrez) 55802
Name mRNA-decapping enzyme 1A Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol DCP1A
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MEALSRAGQE MSLAALKQHD PYITSIADLT GQVALYTFCP KANQWEKTDI EGTLFVYRRS ASPYHGFTIV NRLNMHNLVE PVNKDLEFQL HEPFLLYRNA SLSIYSIWFY DKNDCHRIAK LMADVVEEET RRSQQAARDK QSPSQANGCS DHRPIDILEM LSRAKDEYER NQMGDSNISS PGLQPSTQLS NLGSTETLEE MPSGSQDKSA PSGHKHLTVE ELFGTSLPKE QPAVVGLDSE EMERLPGDAS QKEPNSFLPF PFEQLGGAPQ SETLGVPSAA HHSVQPEITT PVLITPASIT QSNEKHAPTY TIPLSPVLSP TLPAEAPTAQ VPPSLPRNST MMQAVKTTPR QRSPLLNQPV PELSHASLIA NQSPFRAPLN VTNTAGTSLP SVDLLQKLRL TPQHDQIQTQ PLGKGAMVAS FSPAAGQLAT PESFIEPPSK TAAARVAASA SLSNMVLAPL QSMQQNQDPE VFVQPKVLSS AIQVAGAPLV TATTTAVSSV LLAPSVFQQT VTRSSDLERK ASSPSPLTIG TPESQRKPSI ILSKSQLQDT LIHLIKNDSS FLSTLHEVYL QVLTKNKDNH NL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human mRNA-decapping enzyme 1A Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.