missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant M. tuberculosis Signal peptidase I Protein Product Code.: 16059130

enQuireBio™ Recombinant M. tuberculosis Signal peptidase I Protein

Product Code. 16059130
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16059130

Brand: enQuireBio™ QP7516ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Signal peptidase I, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_217419.1
Gene ID (Entrez) 887157
Name M. tuberculosis Signal peptidase I Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol lepB
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity M. tuberculosis
Species E. coli
Protein Tag His-SUMO
Sequence RPYLIPSESM EPTLHGCSTC VGDRIMVDKL SYRFGSPQPG DVIVFRGPPS WNVGYKSIRS HNVAVRWVQN ALSFIGFVPP DENDLVKRVI AVGGQTVQCR SDTGLTVNGR PLKEPYLDPA TMMADPSIYP CLGSEFGPVT VPPGRVWVMG DNRTHSADSR AHCPLLCTDD PLPGTVPVAN VIGKARLIVW PPSRWGVVRS VNPQQGR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant M. tuberculosis Signal peptidase I Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.