missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Lysyl oxidase homolog 4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Lysyl oxidase homolog 4, was expressed in the hosts and tags indicated.
3070.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_444313.3 |
---|---|
Gene ID (Entrez) | 67573 |
Name | Lysyl oxidase homolog 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16021661
|
enQuireBio™
QP7930-ec-10ug |
10 ÎĽg | N/A | ||||||
16061661
|
enQuireBio™
QP7930-ec-50ug |
50 ÎĽg | N/A | ||||||
16011661
|
enQuireBio™
QP7930-ec-100ug |
100 ÎĽg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16041661
|
enQuireBio™
QP7930-ec-200ug |
200 ÎĽg | N/A | ||||||
16051661
|
enQuireBio™
QP7930-ec-500ug |
500 ÎĽg | N/A | ||||||
16031661
|
enQuireBio™
QP7930-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_444313.3 | |
Lysyl oxidase homolog 4 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
67573 | |
Research Use Only | |
Loxl4 | |
Recombinant Protein | |
E. coli | |
QSSGTKKLRL VGPTDRPEEG RLEVLHQGQW GTVCDDDFAL QEATVACRQL GFESALTWAH SAKYGQGEGP IWLDNVRCLG TEKTLDQCGS NGWGVSDCRH SEDVGVVCHP RRQHGYHSEK VSNALGPQGR RLEEVRLKPI LASAKRHSPV TEGAVEVRYD GHWRQVCDQG WTMNNSRVVC GMLGFPSQTS VNSHYYRKVW NLKMKDPKSR LNSLTKKNSF WIHRVDCLGT EPHLAKCQVQ VAPGRGKLRP ACPGGMHAVV SCVAGPHFRR QKPKPTRKES HAEELKVRLR SGAQVGEGRV EVLMNRQWGT VCDHRWNLIS ASVVCRQLGF GSAREALFGA QLGQGLGPIH LSEVRCRGYE RTLGDCLALE GSQNGCQHAN DAAVRCNIPD MGFQNKVRLA GGRNSEEGVV EVQVEVNGVP RWGTVCSDHW GLTEAMVTCR QLGLGFANFA LKDTWYWQGT PEAKEVVMSG VRCSGTEMAL QQCQRHGPVH CSHGPGRFSA GVACMNSAPD LVMNAQLVQE TAYLEDRPLS MLYCAHEENC LSKSADHMDW PYGYRRLLRF SSQIYNLGRA DFRPKAGRHS WIWHQCHRHY HSIEVFTHYD LLTLNGSKVA EGHKASFCLE DTNCPSGVQR RYACANFGEQ GVAVGCWDTY RHDIDCQWVD ITDVGPGDYI FQVVVNPTND VAESDFSNNM IRCRCKYDGQ RVWLHNCHTG DSYRANAELS LEQEQRLRNN LI | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Lysyl oxidase homolog 4 Protein