Learn More
enQuireBio™ Recombinant Mycoplasma Probable cytosol aminopeptidase Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Specificatie
Specificatie
Accession Number | NP_110261.1 |
Gene ID (Entrez) | 876829 |
Name | Mycoplasma Probable cytosol aminopeptidase Protein |
Quantity | 50 μg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | pepA |
Protein Length | Full Length |
Product Type | Recombinant Protein |
Cross Reactivity | Mycoplasma |
Meer weergeven |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
enQuireBio™ Recombinant Mycoplasma Probable cytosol aminopeptidase Protein >
Spot an opportunity for improvement?Share a Content Correction