Learn More
enQuireBio™ Recombinant other Elcatonin Protein
Click to view available options
Quantity:
1 mg
100 mg
200 μg
Unit Size:
100mg
1mg
200µg
Specifications
Specifications
Name | Elcatonin Protein |
Quantity | 100 mg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Biological Activity (based on net peptide) was found to be 6695.2 IU/mg. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Protein Tag | Untagged |
Sequence | Ser-Asn-Leu-Ser-Thr-Asu-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Val-Gly-Ala-Gly-Thr-Pro-NH2 |
Buffer | The protein was lyophilized with no additives. |
Show More |
Product Content Correction
Product Title
enQuireBio™ Recombinant other Elcatonin Protein >
Spot an opportunity for improvement?Share a Content Correction